missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylaniline, 98%

Catalog No. p-7044889
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0862003 1 g
AAL0862006 5 g
AAL0862014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0862003 Supplier Thermo Scientific Chemicals Supplier No. L0862003
Only null left

CAS: 37529-30-9 | C16H27N | 233.399 g/mol

It is used to synthesize p,p'-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize p,p′-Di-n-decyl-azobenzol could be obtained by the reactant of 4-Decylaniline, and the reagents of O2, CuCl, pyridine.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away air and from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

Chemical Identifiers

CAS 37529-30-9
Molecular Formula C16H27N
Molecular Weight (g/mol) 233.399
MDL Number MFCD00007918
InChI Key WGENWPANMZLPIH-UHFFFAOYSA-N
Synonym 4-n-decylaniline, p-decylaniline, benzenamine, 4-decyl, p-n-decylaniline, 4-decylphenylamine, 4-decyphenylamine, 4-decyl-phenylamine, 4-decylaniline, ksc225q8h
PubChem CID 92309
IUPAC Name 4-decylaniline
SMILES CCCCCCCCCCC1=CC=C(C=C1)N

Specifications

Melting Point 23°C to 25°C
Boiling Point 218°C to 220°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.51
Quantity 1 g
Beilstein 2721403
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 233.4
Percent Purity 98%
Chemical Name or Material 4-n-Decylaniline
Show More Show Less
EINECSNumber 253-546-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only