accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 37529-30-9

IUPAC Name: 4-decylaniline
Synonyms: 4-decylaniline
Molecular Weight (g/mol): 233.4
Molecular Formula: C16H27N
InChi Key: WGENWPANMZLPIH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)