missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenyl isocyanate, 98%

Catalog No. AAL0995703
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0995703 1 g
AAL0995714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0995703 Supplier Thermo Scientific Chemicals Supplier No. L0995703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 69342-47-8 | C11H13NO | 175.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69342-47-8
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00013882
InChI Key LJJRXPXDTAUVQU-UHFFFAOYSA-N
Synonym 4-butylphenyl isocyanate, 4-n-butylphenyl isocyanate, benzene, 1-butyl-4-isocyanato, benzene,1-butyl-4-isocyanato, 4-butylbenzenisocyanate, 4-butylphenylisocyanate, acmc-209o7f, 1-butyl-4-isocyanato-benzene, 4-n-butyl phenyl isocyanate
PubChem CID 2796313
IUPAC Name 1-butyl-4-isocyanatobenzene
SMILES CCCCC1=CC=C(C=C1)N=C=O

Specifications

Density 0.992
Boiling Point 107°C to 108°C (3 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.5165
Quantity 1 g
UN Number UN2206
Beilstein 390859
Sensitivity Moisture sensitive
Formula Weight 175.23
Percent Purity 98%
Chemical Name or Material 4-n-Butylphenyl isocyanate
Show More Show Less
Hazard Category H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only