accessibility menu, dialog, popup

UserName

CAS RN 69342-47-8

IUPAC Name: 1-butyl-4-isocyanatobenzene
Synonyms: 1-butyl-4-isocyanatobenzene
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: LJJRXPXDTAUVQU-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)