missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenol, 98%

Catalog No. aaa1281214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1281214 25 g
AAA1281206 5 g
AAA1281222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1281214 Supplier Thermo Scientific Chemicals Supplier No. A1281214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1638-22-8 | C10H14O | 150.221 g/mol

4-n-Butylphenol is used as an Intermediates of liquid Crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Butylphenol is used as an Intermediates of liquid Crystals.

Solubility
Not miscible with water.

Notes
When not in use, tightly seal the container and store in a dry, cool place. Avoid excessive heat and light. Incompatible to oxidizing agents. Stable if stored under proper conditions. Keep container dry. Keep away from heat. Mechanical exhaust required.

Chemical Identifiers

CAS 1638-22-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00041750
InChI Key CYYZDBDROVLTJU-UHFFFAOYSA-N
Synonym 4-n-butylphenol, p-butylphenol, phenol, 4-butyl, p-hydroxybutylbenzene, phenol, p-butyl, 1-hydroxy-4-n-butylbenzene, p-n-butylphenol, 4-butyl-phenol, 4-n-ethylphenol
PubChem CID 15420
ChEBI CHEBI:34437
IUPAC Name 4-butylphenol
SMILES CCCCC1=CC=C(C=C1)O

Specifications

Melting Point ∼22°C
Density 0.98
Boiling Point 245°C
Refractive Index 1.517
Quantity 25 g
UN Number UN3145
Beilstein 2042120
Solubility Information Not miscible with water.
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 4-n-Butylphenol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
EINECSNumber 216-672-5
RTECSNumber SJ8922500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only