missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenol, 98%

Catalog No. p-7081895
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1207606 5 g
AAA1207614 25 g
AAA1207622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1207606 Supplier Thermo Scientific Chemicals Supplier No. A1207606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 122-94-1 | C10H14O2 | 166.22 g/mol

It is an intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate of liquid crystals.

Solubility
Soluble in acetone and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 122-94-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002336
InChI Key MBGGFXOXUIDRJD-UHFFFAOYSA-N
Synonym phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol
PubChem CID 31233
ChEBI CHEBI:34392
IUPAC Name 4-butoxyphenol
SMILES CCCCOC1=CC=C(O)C=C1

Specifications

Melting Point 63°C to 66°C
Quantity 5 g
Solubility Information Soluble in acetone and ethanol.
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-583-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only