missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Butoxybenzonitrile, 97%, Thermo Scientific™
$98.49 - $330.11
Chemical Identifiers
| CAS | 5203-14-5 |
|---|---|
| Molecular Formula | C11H13NO |
| Molecular Weight (g/mol) | 175.231 |
| MDL Number | MFCD00043482 |
| InChI Key | RRGQINKVTNAIBB-UHFFFAOYSA-N |
| Synonym | p-butoxybenzonitrile, 4-n-butoxybenzonitrile, benzonitrile, 4-butoxy, 4-n-butoxy benzonitrile, 4-butoxy-benzonitrile, 4-butoxybenzenecarbonitrile, pubchem7926, acmc-209kxf, 4-butoxybenzonitrile |
| PubChem CID | 138435 |
| IUPAC Name | 4-butoxybenzonitrile |
| SMILES | CCCCOC1=CC=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1657006
|
Thermo Scientific Chemicals
A1657006 |
5 g |
Each for $98.49
|
|
|||||
|
AAA1657014
|
Thermo Scientific Chemicals
A1657014 |
25 g |
Each for $330.11
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5203-14-5 | |
| 175.231 | |
| RRGQINKVTNAIBB-UHFFFAOYSA-N | |
| 138435 | |
| CCCCOC1=CC=C(C=C1)C#N |
| C11H13NO | |
| MFCD00043482 | |
| p-butoxybenzonitrile, 4-n-butoxybenzonitrile, benzonitrile, 4-butoxy, 4-n-butoxy benzonitrile, 4-butoxy-benzonitrile, 4-butoxybenzenecarbonitrile, pubchem7926, acmc-209kxf, 4-butoxybenzonitrile | |
| 4-butoxybenzonitrile |
Specifications
| 5203-14-5 | |
| 242°C to 244°C | |
| Characteristic | |
| 1.5255 | |
| 5 g | |
| p-butoxybenzonitrile, 4-n-butoxybenzonitrile, benzonitrile, 4-butoxy, 4-n-butoxy benzonitrile, 4-butoxy-benzonitrile, 4-butoxybenzenecarbonitrile, pubchem7926, acmc-209kxf, 4-butoxybenzonitrile | |
| CCCCOC1=CC=C(C=C1)C#N | |
| 175.231 | |
| 175.23 | |
| 4-n-Butoxybenzonitrile |
| 0.999 | |
| >110°C (230°F) | |
| C11H13NO | |
| MFCD00043482 | |
| 2834664 | |
| RRGQINKVTNAIBB-UHFFFAOYSA-N | |
| 4-butoxybenzonitrile | |
| 138435 | |
| 97% |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures