missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyaniline, 97%

CAS: 4344-55-2 | C10H15NO | 165.236 g/mol

$70.78 - $229.49

Chemical Identifiers

CAS 4344-55-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00007866
InChI Key UBRIHZOFEJHMIT-UHFFFAOYSA-N
Synonym p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
PubChem CID 20352
IUPAC Name 4-butoxyaniline
SMILES CCCCOC1=CC=C(C=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1752806
View Documents
Thermo Scientific Chemicals
A1752806
5 g
Each for $70.78
Only null left
 
AAA1752814
View Documents
Thermo Scientific Chemicals
A1752814
25 g
Each for $229.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4344-55-2
165.236
UBRIHZOFEJHMIT-UHFFFAOYSA-N
20352
CCCCOC1=CC=C(C=C1)N
C10H15NO
MFCD00007866
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
4-butoxyaniline

Specifications

4344-55-2
84°C to 86°C (0.05 mmHg)
C10H15NO
MFCD00007866
UN2810
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
CCCCOC1=CC=C(C=C1)N
165.236
165.24
4-n-Butoxyaniline
0.992
>110°C (230°F)
1.538
5 g
2084421
UBRIHZOFEJHMIT-UHFFFAOYSA-N
4-butoxyaniline
20352
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 224-402-2

RTECSNumber : BW9420000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only