missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyaniline, 97%

Catalog No. AAA1752814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1752814 25 g
AAA1752806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1752814 Supplier Thermo Scientific Chemicals Supplier No. A1752814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4344-55-2 | C10H15NO | 165.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4344-55-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00007866
InChI Key UBRIHZOFEJHMIT-UHFFFAOYSA-N
Synonym p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
PubChem CID 20352
IUPAC Name 4-butoxyaniline
SMILES CCCCOC1=CC=C(C=C1)N

Specifications

Density 0.992
Boiling Point 84°C to 86°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.538
Quantity 25 g
UN Number UN2810
Beilstein 2084421
Formula Weight 165.24
Percent Purity 97%
Chemical Name or Material 4-n-Butoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 224-402-2
RTECSNumber BW9420000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only