missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinylsulfur trifluoride, 95%

Catalog No. AAL1975114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1975114 25 g
AAL1975103 1 g
AAL1975106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1975114 Supplier Thermo Scientific Chemicals Supplier No. L1975114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51010-74-3 | C4H8F3NOS | 175.169 g/mol

4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing and bases. Store under dry inert gas. It is sensitive to moisture.

Chemical Identifiers

CAS 51010-74-3
Molecular Formula C4H8F3NOS
Molecular Weight (g/mol) 175.169
MDL Number MFCD00037057
InChI Key UFXIRMVZNARBDL-UHFFFAOYSA-N
Synonym morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv
PubChem CID 2733258
IUPAC Name trifluoro(morpholin-4-yl)-$l^{4}-sulfane
SMILES C1COCCN1S(F)(F)F

Specifications

Density 1.436
Boiling Point 41°C to 42°C (0.5 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.4545
Quantity 25 g
UN Number UN3265
Beilstein 606224
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 175.17
Percent Purity 95%
Chemical Name or Material 4-Morpholinylsulfur trifluoride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only