missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Morpholinylcarbonyl Chloride, TCI America™

$58.06 - $145.14

Chemical Identifiers

CAS 15159-40-7
Molecular Formula C5H8ClNO2
Molecular Weight (g/mol) 149.574
MDL Number MFCD00037053
InChI Key XMWFMEYDRNJSOO-UHFFFAOYSA-N
Synonym 4-Chlorocarbonylmorpholine, Morpholinocarbonyl Chloride
PubChem CID 84810
IUPAC Name morpholine-4-carbonyl chloride
SMILES C1COCCN1C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M093525G View Documents TCI America
M093525G
25g Each for $58.06
 
M0935100G View Documents TCI America
M0935100G
100g Each for $145.14
 
Specifications

Chemical Identifiers

15159-40-7
149.574
XMWFMEYDRNJSOO-UHFFFAOYSA-N
84810
C1COCCN1C(=O)Cl
C5H8ClNO2
MFCD00037053
4-Chlorocarbonylmorpholine, Morpholinocarbonyl Chloride
morpholine-4-carbonyl chloride

Specifications

138°C
25g
C5H8ClNO2
3265
XMWFMEYDRNJSOO-UHFFFAOYSA-N
morpholine-4-carbonyl chloride
84810
Clear Liquid at 20°C
4-Morpholinylcarbonyl Chloride
Yellow
15159-40-7
MFCD00037053
4-Chlorocarbonylmorpholine, Morpholinocarbonyl Chloride
C1COCCN1C(=O)Cl
149.574
149.57
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3787

TSCA : Yes