missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Morpholinylcarbonyl Chloride 97.0+%, TCI America™
Supplier: TCI America M0935100G
Specifications
| 4-Morpholinylcarbonyl Chloride | |
| Yellow | |
| C5H8ClNO2 | |
| 100 g | |
| 4-Chlorocarbonylmorpholine, Morpholinocarbonyl Chloride | |
| ClC(=O)N1CCOCC1 | |
| 149.57 | |
| 149.57 | |
| Liquid |
| 15159-40-7 | |
| 138°C | |
| MFCD00037053 | |
| 3265 | |
| XMWFMEYDRNJSOO-UHFFFAOYSA-N | |
| morpholine-4-carbonyl chloride | |
| 84810 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 15159-40-7 | |
| 149.57 | |
| XMWFMEYDRNJSOO-UHFFFAOYSA-N | |
| 84810 | |
| ClC(=O)N1CCOCC1 |
| C5H8ClNO2 | |
| MFCD00037053 | |
| 4-Chlorocarbonylmorpholine, Morpholinocarbonyl Chloride | |
| morpholine-4-carbonyl chloride |
Safety and Handling
EINECSNumber : (5)-3787
TSCA : Yes