missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC29304EA
Change view
Click to view available options
Quantity:
250 mg
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29304EA 10 g Amber Glass Bottle
CC29304CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC29304EA Supplier Thermo Scientific Chemicals Supplier No. CC29304EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82413-63-6
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD05865107
InChI Key KMAHWHPUXGNVBN-UHFFFAOYSA-N
Synonym 4-morpholinomethyl benzaldehyde, 4-morpholin-4-ylmethyl benzaldehyde, 4-morpholinomethyl-benzaldehyde, 4-4-morpholinylmethyl benzaldehyde, 4-morpholin-4-ylmethyl-benzaldehyde, 4-4-carboxaldehydebenzyl morpholine, benzaldehyde,4-4-morpholinylmethyl
PubChem CID 2795498
IUPAC Name 4-(morpholin-4-ylmethyl)benzaldehyde
SMILES C1COCCN1CC2=CC=C(C=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)benzaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.