missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinoaniline 98.0+%, TCI America™

Catalog No. p-7132752
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14661G 1 g
M14665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14661G Supplier TCI America Supplier No. M14661G
Only null left

Chemical Identifiers

CAS 2524-67-6
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00006169
InChI Key PHNDZBFLOPIMSM-UHFFFAOYSA-N
Synonym 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
PubChem CID 75655
IUPAC Name 4-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=C(C=C2)N

Specifications

Melting Point 134°C
Quantity 1 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Morpholinoaniline
EINECSNumber (5)-0870
TSCA Yes