accessibility menu, dialog, popup

UserName

CAS RN 2524-67-6

IUPAC Name: 4-(morpholin-4-yl)aniline
Synonyms: 4-(morpholin-4-yl)aniline
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: PHNDZBFLOPIMSM-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1CCOCC1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)