Learn More
4-Morpholinoaniline, 97%, Thermo Scientific™
$157.94 - $157.94
Chemical Identifiers
| CAS | 2524-67-6 |
|---|---|
| Molecular Formula | C10H14N2O |
| Molecular Weight (g/mol) | 178.235 |
| MDL Number | MFCD00006169 |
| InChI Key | PHNDZBFLOPIMSM-UHFFFAOYSA-N |
| Synonym | 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine |
| PubChem CID | 75655 |
| IUPAC Name | 4-morpholin-4-ylaniline |
| SMILES | C1COCCN1C2=CC=C(C=C2)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BTB01373EB
|
Thermo Scientific Chemicals
BTB01373EB |
25 g | Amber Glass Bottle |
Each for $157.94
|
|
||||
Chemical Identifiers
| 2524-67-6 | |
| 178.235 | |
| PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
| 75655 | |
| C1COCCN1C2=CC=C(C=C2)N |
| C10H14N2O | |
| MFCD00006169 | |
| 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
| 4-morpholin-4-ylaniline |
Specifications
| 2524-67-6 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00006169 | |
| 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
| C1COCCN1C2=CC=C(C=C2)N | |
| 178.235 | |
| 178.23 | |
| 4-Morpholinoaniline |
| 96.5 | |
| 97% | |
| C10H14N2O | |
| 25 g | |
| PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
| 4-morpholin-4-ylaniline | |
| 75655 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning