missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylvaleryl Chloride 98.0+%, TCI America™
Supplier: TCI America M228725ML
Specifications
| 4-Methylvaleryl Chloride | |
| Yellow | |
| C6H11ClO | |
| 25 mL | |
| 4-Methylvaleroyl Chloride | |
| CC(C)CCC(Cl)=O | |
| 134.60 | |
| 134.60 | |
| Reagent |
| 38136-29-7 | |
| 144°C | |
| MFCD00018814 | |
| 2920 | |
| SVWCVXFHTHCJJB-UHFFFAOYSA-N | |
| 4-methylpentanoyl chloride | |
| 170008 | |
| ≥98.0% (T) | |
| Liquid |
Chemical Identifiers
| 38136-29-7 | |
| 134.60 | |
| SVWCVXFHTHCJJB-UHFFFAOYSA-N | |
| 170008 | |
| CC(C)CCC(Cl)=O |
| C6H11ClO | |
| MFCD00018814 | |
| 4-Methylvaleroyl Chloride | |
| 4-methylpentanoyl chloride |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes