accessibility menu, dialog, popup

UserName

H3CCH3ClO

CAS RN 38136-29-7

IUPAC Name: 4-methylpentanoyl chloride
Synonyms: 4-methylpentanoyl chloride
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: SVWCVXFHTHCJJB-UHFFFAOYSA-N
SMILES: CC(C)CCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results