missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl sulfate potassium salt, 98%

Catalog No. p-7043198
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66160ME 500 mg
AAJ6616003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66160ME Supplier Thermo Scientific Chemicals Supplier No. J66160ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for sulfatase | CAS: 15220-11-8 | C10H7KO6S | 294.318 g/mol

4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

Solubility
Soluble in water at 5mg/ml

Notes
Excitation wavelength, 334 nm; emission wavelength, 370nm (pH 10.4). Reaction product: excitiation wavelength, 360nm; emission wavelength, 449nm

Chemical Identifiers

CAS 15220-11-8
Molecular Formula C10H7KO6S
Molecular Weight (g/mol) 294.318
MDL Number MFCD00016970
InChI Key CSOCSPXOODWGLJ-UHFFFAOYSA-M
Synonym 4-methylumbelliferyl sulfate potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt, 4-methylumbelliferyl sulfate, potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt 1:1, potassium 4-methyl-2-oxo-2h-chromen-7-yl sulfate, potassium 4-methyl-2-oxochromen-7-yl sulfate, c10h7o6s.k, potassium 4-methyl-2-oxo-2h-1-benzopyran-7-yl sulphate, potassium 4-methylumbelliferyl sulfate
PubChem CID 5044226
IUPAC Name potassium;(4-methyl-2-oxochromen-7-yl) sulfate
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OS(=O)(=O)[O-].[K+]

Specifications

Color White to Yellow
Quantity 500 mg
Beilstein 3803203
Solubility Information Soluble in water at 5mg/ml
Formula Weight 294.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl sulfate potassium salt
EINECSNumber 239-272-2
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only