accessibility menu, dialog, popup

UserName

KCH3OOOSOOO

CAS RN 15220-11-8

IUPAC Name: potassium 4-methyl-2-oxo-2H-chromen-7-yl sulfate
Synonyms: potassium 4-methyl-2-oxochromen-7-yl sulfate
Molecular Weight (g/mol): 294.32
Molecular Formula: C10H7KO6S
InChi Key: CSOCSPXOODWGLJ-UHFFFAOYSA-M
SMILES: [K+].CC1=CC(=O)OC2=CC(OS([O-])(=O)=O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results