missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl alpha-L-Arabinopyranoside 98.0+%, TCI America™

Catalog No. p-7133539
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M302550MG 50 mg
M3025250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M302550MG Supplier TCI America Supplier No. M302550MG
Only null left

Chemical Identifiers

CAS 69414-26-2
Molecular Formula C15H16O7
Molecular Weight (g/mol) 308.286
MDL Number MFCD00064087
InChI Key JWIYLOHVJDJZOQ-HPEDKQMDSA-N
PubChem CID 18634901
IUPAC Name 4-methyl-7-[(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(CO3)O)O)O

Specifications

Color White
Quantity 50 mg
Formula Weight 308.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl alpha-L-Arabinopyranoside
TSCA No
Recommended Storage Freezer (-20 °C)