accessibility menu, dialog, popup

UserName

CAS RN 69414-26-2

IUPAC Name: 4-methyl-7-[(3,4,5-trihydroxyoxan-2-yl)oxy]-2H-chromen-2-one
Synonyms: 4-methyl-7-[(3,4,5-trihydroxyoxan-2-yl)oxy]chromen-2-one
Molecular Weight (g/mol): 308.29
Molecular Formula: C15H16O7
InChi Key: JWIYLOHVJDJZOQ-UHFFFAOYNA-N
SMILES: CC1=CC(=O)OC2=CC(OC3OCC(O)C(O)C3O)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)