missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside 98.0+%, TCI America™

Supplier:  TCI America M30291G

Quantity 1 g

 View more versions of this product

Catalog No. M30291G


Only null left

Description

Description

Specifications

Chemical Identifiers

36476-29-6
378.357
4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide
N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
C18H20NO8-
QCTHLCFVVACBSA-SOVHRIKKSA-M
22842632
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]

Specifications

4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside
207°C
C18H20NO8-
4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]
378.357
379.37
Crystalline Powder
36476-29-6
White-Yellow
1 g
QCTHLCFVVACBSA-SOVHRIKKSA-M
N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
22842632
≥98.0% (HPLC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
Documents

Documents

Promotions

Promotions