missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. p-7133543
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3029250MG 250 mg
M30291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3029250MG Supplier TCI America Supplier No. M3029250MG
Only null left

Chemical Identifiers

CAS 36476-29-6
Molecular Formula C18H20NO8-
Molecular Weight (g/mol) 378.357
InChI Key QCTHLCFVVACBSA-SOVHRIKKSA-M
Synonym 4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide
PubChem CID 22842632
IUPAC Name N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 250 mg
Formula Weight 379.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer (-20 °C)