missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)toluene 98.0+%, TCI America™

Catalog No. M094925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M094925G 25 g
M094910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M094925G Supplier TCI America Supplier No. M094925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-13-2
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00008560
InChI Key VHILIAIEEYLJNA-UHFFFAOYSA-N
Synonym methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
PubChem CID 69334
IUPAC Name 1-methyl-4-(methylsulfanyl)benzene
SMILES CSC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 25 g
Formula Weight 138.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Methylthio)toluene