missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenylacetic acid, 97%

Catalog No. AAH2688806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2688806 5 g
AAH2688803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2688806 Supplier Thermo Scientific Chemicals Supplier No. H2688806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16188-55-9 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16188-55-9
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00192325
InChI Key AHMLFHMRRBJCRM-UHFFFAOYSA-N
Synonym 4-methylthio phenylacetic acid, 4-methylthiophenylacetic acid, 2-4-methylthio phenyl acetic acid, 4-methylsulfanylphenyl acetic acid, benzeneacetic acid, 4-methylthio, 4-methylsulfanyl-phenyl-acetic acid, 2-4-methylsulfanyl phenyl acetic acid, 4-methylsulfanyl phenyl acetic acid, 2-4-methylthiophenyl acetic acid, 4-methylthiophenylaceticacid
PubChem CID 4983912
IUPAC Name 2-(4-methylsulfanylphenyl)acetic acid
SMILES CSC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)phenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only