missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenyl isothiocyanate, 97%

Catalog No. AAL0927006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0927006 5 g
AAL0927003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0927006 Supplier Thermo Scientific Chemicals Supplier No. L0927006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 15863-41-9 | C8H7NS2 | 181.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15863-41-9
Molecular Formula C8H7NS2
Molecular Weight (g/mol) 181.271
MDL Number MFCD00041368
InChI Key CEBAJHCAFXYWNT-UHFFFAOYSA-N
Synonym 4-methylthio phenyl isothiocyanate, 4-methylsulfanyl phenyl isothiocyanate, 1-isothiocyanato-4-methylsulfanyl benzene, benzene,1-isothiocyanato-4-methylthio, acmc-20amu3, 4-methylthiophenylisothiocyanate, 4-methylthio phenylisothiocyanate, 1-isothiocyanato-4-methylthio-benzene, 1-isothiocyanato-4-methylsulfanyl benzene #
PubChem CID 519185
IUPAC Name 1-isothiocyanato-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 48°C to 51°C
Boiling Point 127°C to 128°C (1 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2082547
Sensitivity Moisture sensitive
Formula Weight 181.28
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)phenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only