missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenol 98.0+%, TCI America™

Catalog No. p-7132173
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
M044125G 25 g
M0441250G 250 g
2 options

Catalog No. M044125G

Supplier: TCI America M044125G

Only null left

Chemical Identifiers

CAS 1073-72-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002351
InChI Key QASBCTGZKABPKX-UHFFFAOYSA-N
Synonym 4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
PubChem CID 14086
ChEBI CHEBI:38862
IUPAC Name 4-(methylsulfanyl)phenol
SMILES CSC1=CC=C(O)C=C1

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)phenol
EINECSNumber (3)-1099
RTECSNumber SM1578500
TSCA Yes