missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenol, 98%

Catalog No. AAH2622509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2622509 10 g
AAH2622518 50 g
AAH2622530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2622509 Supplier Thermo Scientific Chemicals Supplier No. H2622509

CAS: 1073-72-9 | C7H8OS | 140.20 g/mol

4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water 9.59 g/ l.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1073-72-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002351
InChI Key QASBCTGZKABPKX-UHFFFAOYSA-N
Synonym 4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
PubChem CID 14086
ChEBI CHEBI:38862
IUPAC Name 4-methylsulfanylphenol
SMILES CSC1=CC=C(O)C=C1

Specifications

Melting Point 85°C to 87°C
Boiling Point 153°C to 156°C (20 mmHg)
Odor Stench
Quantity 10 g
Solubility Information Soluble in water 9.59g/ l.
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)phenol
Hazard Category H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
EINECSNumber 214-031-4
RTECSNumber SM1578500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only