missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-m-cresol 98.0+%, TCI America™

Catalog No. M053125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M053125G Supplier TCI America Supplier No. M053125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3120-74-9
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00045773
InChI Key VKALYYFVKBXHTF-UHFFFAOYSA-N
Synonym 3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
PubChem CID 18391
ChEBI CHEBI:38681
IUPAC Name 3-methyl-4-methylsulfanylphenol
SMILES CC1=C(C=CC(=C1)O)SC

Specifications

Melting Point 54°C
Color Red-White
Boiling Point 151°C
Quantity 25 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)-m-cresol
EINECSNumber (3)-1100
RTECSNumber GP2225000
TSCA Yes