missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)imidazole, 97%

Catalog No. AAH63990MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63990MD 250 mg
AAH6399003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63990MD Supplier Thermo Scientific Chemicals Supplier No. H63990MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83553-60-0 | C4H6N2S | 114.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83553-60-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD03844702
InChI Key NLIJIXLRIYPOHM-UHFFFAOYSA-N
Synonym 4-methylthio-1h-imidazole, 4-methylthio imidazole, 5-methylthio-1h-imidazole, 4-methylsulfanyl-1h-imidazole, pubchem14136, 4-methylthio ;-1h-imidazole, 5-methylsulfanyl-1h-imidazole, 1h-imidazole,4-methylthio-9ci, 1h-imidazole, 4-methylthio
PubChem CID 13084405
IUPAC Name 5-methylsulfanyl-1H-imidazole
SMILES CSC1=CN=CN1

Specifications

Quantity 250 mg
Formula Weight 114.17
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only