missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)butanol, 99%

Catalog No. p-7026599
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2267303 1 g
AAB2267306 5 g
AAB2267314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2267303 Supplier Thermo Scientific Chemicals Supplier No. B2267303

CAS: 20582-85-8 | C5H12OS | 120.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20582-85-8
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00002973
InChI Key JNTVUHZXIJFHAU-UHFFFAOYSA-N
Synonym 4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound
PubChem CID 519793
SMILES CSCCCCO

Specifications

Density 0.999
Boiling Point 95°C to 98°C (20 mmHg)
Flash Point 101°C (215°F)
Odor Unpleasant
Refractive Index 1.487
Linear Formula CH3S(CH2)4OH
Quantity 1 g
IUPAC Name 4-methylsulfanylbutan-1-ol
Formula Weight 120.21
Percent Purity 99%
Chemical Name or Material 4-(Methylthio)butanol
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only