missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzoic acid, 97%

Catalog No. AAB2570603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2570603 1 g
AAB2570606 5 g
AAB2570614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2570603 Supplier Thermo Scientific Chemicals Supplier No. B2570603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13205-48-6 | C8H8O2S | 168.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13205-48-6
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00002555
InChI Key KWHCPERWLHBLOT-UHFFFAOYSA-N
Synonym 4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa
PubChem CID 83230
IUPAC Name 4-methylsulfanylbenzoic acid
SMILES CSC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Beilstein 2082809
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzoic acid
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-171-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only