missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzaldehyde 97.0+%, TCI America™

Catalog No. p-7132333
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M073925G 25 g
M0739250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M073925G Supplier TCI America Supplier No. M073925G
Only null left

Chemical Identifiers

CAS 3446-89-7
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00006948
InChI Key QRVYABWJVXXOTN-UHFFFAOYSA-N
Synonym 4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
PubChem CID 76985
IUPAC Name 4-methylsulfanylbenzaldehyde
SMILES CSC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
Formula Weight 152.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Methylthio)benzaldehyde