missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-(Methylthio)acetophenone, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 80°C to 82°C |
| Boiling Point | 135°C (1 mmHg) |
| Quantity | 5 g |
| Beilstein | 1860517 |
| Formula Weight | 166.24 |
| Percent Purity | 99% |
| Chemical Name or Material | 4'-(Methylthio)acetophenone |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 217-213-1 |
| TSCA | No |
RUO – Research Use Only