missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylthio)acetophenone, 99%

Catalog No. AAL1807406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1807406 5 g
AAL1807414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1807406 Supplier Thermo Scientific Chemicals Supplier No. L1807406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1778-09-2 | C9H10OS | 166.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1778-09-2
Molecular Formula C9H10OS
Molecular Weight (g/mol) 166.24
MDL Number MFCD00039835
InChI Key JECUZQLBQKNEMW-UHFFFAOYSA-N
Synonym 4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone
PubChem CID 74501
IUPAC Name 1-(4-methylsulfanylphenyl)ethanone
SMILES CSC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 80°C to 82°C
Boiling Point 135°C (1 mmHg)
Quantity 5 g
Beilstein 1860517
Formula Weight 166.24
Percent Purity 99%
Chemical Name or Material 4'-(Methylthio)acetophenone
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-213-1
TSCA No

RUO – Research Use Only