missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthio-2-butanone 97.0+%, TCI America™

Catalog No. p-7132979
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20315G 5 g
M203125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20315G Supplier TCI America Supplier No. M20315G

Chemical Identifiers

CAS 34047-39-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00040029
InChI Key DRGHCRKOWMAZAO-UHFFFAOYSA-N
Synonym 4-methylthio-2-butanone, 2-butanone, 4-methylthio, 4-methylthio butan-2-one, unii-5703w1o1ab, 4-methylsulfanyl butan-2-one, potato butanone, fema no. 3375, 5-thiahexane-2-one, acmc-1afub
PubChem CID 61922
IUPAC Name 4-methylsulfanylbutan-2-one
SMILES CC(=O)CCSC

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
Formula Weight 118.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylthio-2-butanone