missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7132918
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M19325G 5 g
M193225G 25 g
2 options

Catalog No. M19325G

Supplier: TCI America M19325G

Only null left

Chemical Identifiers

CAS 4052-30-6
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00007564
InChI Key AJBWNNKDUMXZLM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzoic acid, p-methylsulfonylbenzoic acid, 4-methanesulfonyl-benzoic acid, 4-methylsulphonylbenzoic acid, 4-methylsulphonyl benzoic acid, 4-methanesulfonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulphonyl benzoic acid, 4-methanesulfonyl benzoic acid
PubChem CID 77670
IUPAC Name 4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 265°C
Color White
Quantity 5 g
Formula Weight 200.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)benzoic Acid