missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzoic acid, 98%

Catalog No. AAH5023003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5023003 1 g
AAH5023006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5023003 Supplier Thermo Scientific Chemicals Supplier No. H5023003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4052-30-6 | C8H8O4S | 200.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4052-30-6
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00007564
InChI Key AJBWNNKDUMXZLM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzoic acid, p-methylsulfonylbenzoic acid, 4-methanesulfonyl-benzoic acid, 4-methylsulphonylbenzoic acid, 4-methylsulphonyl benzoic acid, 4-methanesulfonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulphonyl benzoic acid, 4-methanesulfonyl benzoic acid
PubChem CID 77670
IUPAC Name 4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 270°C to 275°C
Quantity 1 g
Formula Weight 200.21
Percent Purity 98%
Chemical Name or Material 4-(Methylsulfonyl)benzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 223-765-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only