missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%

Catalog No. p-7033207
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5026403 1 g
AAH5026406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5026403 Supplier Thermo Scientific Chemicals Supplier No. H5026403
Only null left

CAS: 82964-91-8 | C7H7ClO4S2 | 254.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82964-91-8
Molecular Formula C7H7ClO4S2
Molecular Weight (g/mol) 254.70
MDL Number MFCD00216486
InChI Key TYJOQICPGZGYDT-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzenesulfonyl chloride, 4-methylsulfonyl benzene-1-sulfonyl chloride, 4-methanesulfonylbenzene-1-sulfonyl chloride, 4-methylsulphonyl benzenesulphonyl chloride, 4-methanesulfonylbenzenesulfonyl chloride, 4-methylsulfonylbenzenesulfonylchloride, pubchem5614, acmc-20amd2, chloro 4-methylsulfonyl phenyl sulfone, 4-methylsulfonyl phenylsulfonyl chloride
PubChem CID 2735182
IUPAC Name 4-methylsulfonylbenzenesulfonyl chloride
SMILES CS(=O)(=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 158°C to 163°C
Quantity 1 g
UN Number UN3261
Formula Weight 254.71
Percent Purity 95%
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only