missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)aniline 98.0+%, TCI America™

Catalog No. A272525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A272525G 25 g
A2725100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A272525G Supplier TCI America Supplier No. A272525G

Chemical Identifiers

CAS 5470-49-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025353
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
Synonym 4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
PubChem CID 79617
IUPAC Name 4-methanesulfonylaniline
SMILES CS(=O)(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 137°C
Color White
Quantity 25 g
Formula Weight 171.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)aniline
EINECSNumber (3)-2137
TSCA No