Learn More
4-(Methylsulfinyl)benzeneboronic acid, 98%
CAS: 166386-48-7 | C7H9BO3S | 184.02 g/mol
Supplier: Thermo Scientific Chemicals L17865MC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-(Methylsulfinyl)benzeneboronic acid | |
| ∼170°C | |
| MFCD02093071 | |
| 4-methanesulfinyl benzeneboronic acid, 4-methylsulfinyl phenyl boronic acid, 4-methylsulphinyl benzeneboronic acid, 4-methylsulfinylphenyl boronic acid, 4-methanesulphinylphenylboronic acid, 4-methylsulfinyl phenylboronic acid, boronic acid, 4-methylsulfinyl phenyl, 4-boronophenyl methyl sulfoxide, 4-methanesulfinylphenylboronic acid | |
| CS(=O)C1=CC=C(C=C1)B(O)O | |
| 184.02 | |
| 184.02 |
| 166386-48-7 | |
| C7H9BO3S | |
| 100 mg | |
| YOTGALZTDVXUKZ-UHFFFAOYNA-N | |
| (4-methylsulfinylphenyl)boronic acid | |
| 2773531 | |
| 98% |
Chemical Identifiers
| 166386-48-7 | |
| 184.02 | |
| YOTGALZTDVXUKZ-UHFFFAOYNA-N | |
| 2773531 | |
| CS(=O)C1=CC=C(C=C1)B(O)O |
| C7H9BO3S | |
| MFCD02093071 | |
| 4-methanesulfinyl benzeneboronic acid, 4-methylsulfinyl phenyl boronic acid, 4-methylsulphinyl benzeneboronic acid, 4-methylsulfinylphenyl boronic acid, 4-methanesulphinylphenylboronic acid, 4-methylsulfinyl phenylboronic acid, boronic acid, 4-methylsulfinyl phenyl, 4-boronophenyl methyl sulfoxide, 4-methanesulfinylphenylboronic acid | |
| (4-methylsulfinylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only