Learn More
4-Methylsulfamoylbenzeneboronic acid, 97%
CAS: 226396-31-2 | C7H10BNO4S | 215.03 g/mol
$213.60 - $670.82
Chemical Identifiers
| CAS | 226396-31-2 |
|---|---|
| Molecular Formula | C7H10BNO4S |
| Molecular Weight (g/mol) | 215.03 |
| MDL Number | MFCD06659864 |
| InChI Key | DOQOQZHSIBBHMY-UHFFFAOYSA-N |
| Synonym | 4-n-methylsulfamoyl phenyl boronic acid, methyl 4-boronobenzenesulfonamide, 4-methylsulfamoyl phenyl boronic acid, 4-methylaminosulphonyl benzene boronic acid, 4-n-methylsulfamoyl phenylboronic acid, 4-methylsulfamoyl phenylboronic acid, 4-methylsulfamoylphenylboronic acid, boronic acid, 4-methylamino sulfonyl phenyl, acmc-209fxs |
| PubChem CID | 44118763 |
| IUPAC Name | [4-(methylsulfamoyl)phenyl]boronic acid |
| SMILES | CNS(=O)(=O)C1=CC=C(C=C1)B(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52493MD
|
Thermo Scientific Chemicals
H52493MD |
250 mg |
Each for $213.60
|
|
|||||
|
AAH5249303
|
Thermo Scientific Chemicals
H5249303 |
1 g |
Each for $670.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 226396-31-2 | |
| 215.03 | |
| DOQOQZHSIBBHMY-UHFFFAOYSA-N | |
| 44118763 | |
| CNS(=O)(=O)C1=CC=C(C=C1)B(O)O |
| C7H10BNO4S | |
| MFCD06659864 | |
| 4-n-methylsulfamoyl phenyl boronic acid, methyl 4-boronobenzenesulfonamide, 4-methylsulfamoyl phenyl boronic acid, 4-methylaminosulphonyl benzene boronic acid, 4-n-methylsulfamoyl phenylboronic acid, 4-methylsulfamoyl phenylboronic acid, 4-methylsulfamoylphenylboronic acid, boronic acid, 4-methylamino sulfonyl phenyl, acmc-209fxs | |
| [4-(methylsulfamoyl)phenyl]boronic acid |
Specifications
| 226396-31-2 | |
| C7H10BNO4S | |
| 250 mg | |
| DOQOQZHSIBBHMY-UHFFFAOYSA-N | |
| [4-(methylsulfamoyl)phenyl]boronic acid | |
| 44118763 | |
| 97% |
| 136°C to 140°C | |
| MFCD06659864 | |
| 4-n-methylsulfamoyl phenyl boronic acid, methyl 4-boronobenzenesulfonamide, 4-methylsulfamoyl phenyl boronic acid, 4-methylaminosulphonyl benzene boronic acid, 4-n-methylsulfamoyl phenylboronic acid, 4-methylsulfamoyl phenylboronic acid, 4-methylsulfamoylphenylboronic acid, boronic acid, 4-methylamino sulfonyl phenyl, acmc-209fxs | |
| CNS(=O)(=O)C1=CC=C(C=C1)B(O)O | |
| 215.03 | |
| 215.03 | |
| 4-Methylsulfamoylbenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only