missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsalicylic acid, 99%

Catalog No. AAL0336722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0336722 100 g
AAL0336714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0336722 Supplier Thermo Scientific Chemicals Supplier No. L0336722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-85-1 | C8H8O3 | 152.149 g/mol

4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

Solubility
Soluble in water 10 g/L @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 50-85-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002454
InChI Key NJESAXZANHETJV-UHFFFAOYSA-N
Synonym 4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
PubChem CID 5788
ChEBI CHEBI:20450
IUPAC Name 2-hydroxy-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)O

Specifications

Melting Point 176°C to 180°C
Odor Odorless
Quantity 100 g
Beilstein 2208140
Merck Index 14,2580
Solubility Information Soluble in water 10g/L @ 20?C.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 4-Methylsalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-068-3
RTECSNumber GP3920100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only