missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsalicylic acid, 98%

CAS: 50-85-1 | C8H8O3 | 152.149 g/mol

$309.05 - $309.05

Chemical Identifiers

CAS 50-85-1
MDL Number MFCD00002454
InChI Key NJESAXZANHETJV-UHFFFAOYSA-N
Synonym 4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
PubChem CID 5788
ChEBI CHEBI:20450
IUPAC Name 2-hydroxy-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223541000
SDS View Documents
Thermo Scientific Chemicals
223541000
100 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-85-1
NJESAXZANHETJV-UHFFFAOYSA-N
5788
2-hydroxy-4-methylbenzoic acid
MFCD00002454
4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
CHEBI:20450
CC1=CC(=C(C=C1)C(=O)O)O

Specifications

50-85-1
100.0
Beige-Gray to White
98%
C8H8O3
MFCD00002454
10, 233
4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
CC1=CC(=C(C=C1)C(=O)O)O
152.149
CHEBI:20450
98%
4-Methylsalicylic acid
97.5
175.0°C to 179.0°C
Authentic
Glass bottle
CH3C6H3(OH)CO2H
100 g
15, 2569
NJESAXZANHETJV-UHFFFAOYSA-N
2-hydroxy-4-methylbenzoic acid
5788
152.15
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only