missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyrimidine, 98%

Catalog No. AAA1317006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1317006 5 g
AAA1317014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1317006 Supplier Thermo Scientific Chemicals Supplier No. A1317006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3438-46-8 | C5H6N2 | 94.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3438-46-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006115
InChI Key LVILGAOSPDLNRM-UHFFFAOYSA-N
Synonym pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci
PubChem CID 18922
IUPAC Name 4-methylpyrimidine
SMILES CC1=NC=NC=C1

Specifications

Density 1.03
Boiling Point 141°C to 142°C
Flash Point 40°C (104°F)
Refractive Index 1.496
Quantity 5 g
UN Number UN1993
Beilstein 105781
Formula Weight 94.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylpyrimidine
Show More Show Less
Hazard Category H226-H360D
Hazard Statement GHS H Statement

H226

Flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 222-344-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only