missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyridine N-Oxide 98.0+%, TCI America™

Catalog No. P042025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P042025G Supplier TCI America Supplier No. P042025G

Chemical Identifiers

CAS 1003-67-4
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006210
InChI Key IWYYIZOHWPCALJ-UHFFFAOYSA-N
Synonym 4-picoline-n-oxide, 4-picoline n-oxide, 4-methylpyridine n-oxide, 4-methylpyridine 1-oxide, pyridine, 4-methyl-, 1-oxide, 4-picoline 1-oxide, 4-picoline, 1-oxide, gamma-picoline n-oxide, gamma-picoline 1-oxide, 4-methylpyridin-1-ium-1-olate
PubChem CID 13857
IUPAC Name 4-methylpyridin-1-ium-1-olate
SMILES CC1=CC=[N+]([O-])C=C1

Specifications

Melting Point 184°C
Quantity 25 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylpyridine N-Oxide
RTECSNumber UT5800000
TSCA Yes