missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenoxyacetic Acid 98.0+%, TCI America™

Catalog No. p-7133394
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M27295G 5 g
M272925G 25 g
2 options

Catalog No. M27295G

Supplier: TCI America M27295G

Only null left

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylphenoxyacetic Acid