missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenoxyacetic acid, 98%

Catalog No. AAA1323030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323030 250 g
AAA1323009 10 g
AAA1323018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1323030 Supplier Thermo Scientific Chemicals Supplier No. A1323030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 940-64-7 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O

Specifications

Melting Point 137°C to 142°C
Quantity 250 g
Beilstein 2047591
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4-Methylphenoxyacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-374-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only