missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyloxazole-5-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M26631G
Specifications
| 4-Methyloxazole-5-carboxylic Acid | |
| 242°C | |
| MFCD01571070 | |
| 4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl | |
| CC1=C(OC=N1)C(=O)O | |
| 127.099 | |
| 127.10 | |
| Crystalline Powder |
| 2510-32-9 | |
| C5H5NO3 | |
| 1 g | |
| ZIXUNDOOBLSXPE-UHFFFAOYSA-N | |
| 4-methyl-1,3-oxazole-5-carboxylic acid | |
| 292311 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2510-32-9 | |
| 127.099 | |
| ZIXUNDOOBLSXPE-UHFFFAOYSA-N | |
| 292311 | |
| CC1=C(OC=N1)C(=O)O |
| C5H5NO3 | |
| MFCD01571070 | |
| 4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl | |
| 4-methyl-1,3-oxazole-5-carboxylic acid |
Safety and Handling
TSCA : No